CH4182076994 ChemBridge Corporation

3-(4-fluorophenyl)tetrahydro-3-furancarbaldehyde

  • MDL number MFCD11053963
  • Cas Number : 1060817-44-8
  • Molecular Weight : 194.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4029252

Safety Information

MSDS https://www.hit2lead.com/sds/4029252_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart