CH4182077017 ChemBridge Corporation

5-(chloromethyl)-4-isopropyl-1,2,3-thiadiazole

  • MDL number MFCD11053980
  • Cas Number : 1060817-54-0
  • Molecular Weight : 176.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/4030838

Safety Information

msds https://www.hit2lead.com/building-blocks/4030838
Availability details for
Quantity
  • Availability
  • Add to Cart