CH4182077054 ChemBridge Corporation

1-(3-ethyl-5-isoxazolyl)-N-methylmethanamine

  • MDL number MFCD11054022
  • Cas Number : 942519-63-3
  • Molecular Weight : 140.183

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4034176

Safety Information

MSDS https://www.hit2lead.com/sds/4034176_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart