CH4182077077 ChemBridge Corporation

2-(4-bromophenyl)-6-methyl-4-pyrimidinol

  • MDL number MFCD00697484
  • Cas Number : 180606-52-4
  • Molecular Weight : 265.106

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5339492

Safety Information

MSDS https://www.hit2lead.com/sds/5339492_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart