CH4182077114 ChemBridge Corporation

4-bromo-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD00463984
  • Cas Number : 13745-17-0
  • Molecular Weight : 190.9828

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4029787

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4029787
Availability details for
Quantity
  • Availability
  • Add to Cart