CH4182077163 ChemBridge Corporation

(5-bromo-1H-benzimidazol-2-yl)methanol

  • MDL number MFCD03197450
  • Cas Number : 540516-28-7
  • Molecular Weight : 227.058

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4030161

Safety Information

MSDS https://www.hit2lead.com/sds/4030161_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart