CH4182077178 ChemBridge Corporation

3,4-dihydro-1(2H)-quinolinyl(oxo)acetic acid

  • MDL number MFCD10018451
  • Cas Number : 1018565-99-5
  • Molecular Weight : 205.2099

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Assay 0.95
Storage temp. Room Temperature

Safety Information

msds https://www.hit2lead.com/building-blocks/4030227
Availability details for
Quantity
  • Availability
  • Add to Cart