CH4182077199 ChemBridge Corporation

N-cyclopropyltetrahydro-2H-pyran-4-amine

  • MDL number MFCD06658420
  • Cas Number : 211814-16-3
  • Molecular Weight : 141.2108

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4030385

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4030385
Availability details for
Quantity
  • Availability
  • Add to Cart