CH4182077220 ChemBridge Corporation

3-(4-methyl-1-piperidinyl)-1-propanamine

  • MDL number MFCD00129008
  • Cas Number : 6241-30-1
  • Molecular Weight : 156.2685

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4030577

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4030577
Availability details for
Quantity
  • Availability
  • Add to Cart