CH4182077225 ChemBridge Corporation

oxo(1-piperidinyl)acetic acid

  • MDL number MFCD06740188
  • Cas Number : 4706-33-6
  • Molecular Weight : 157.1671

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4030610

Safety Information

MSDS https://www.hit2lead.com/sds/4030610_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart