CH4182077251 ChemBridge Corporation

6-amino-2,4-dimethyl-2H-1,4-benzoxazin-3(4H)-one

  • MDL number MFCD07788350
  • Cas Number : 70488-69-6
  • Molecular Weight : 192.2145

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4030801

Safety Information

MSDS https://www.hit2lead.com/sds/4030801_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart