CH4182077293 ChemBridge Corporation

3-(chlorosulfonyl)-4-methylbenzoic acid

  • MDL number MFCD00625710
  • Cas Number : 2548-29-0
  • Molecular Weight : 234.6568

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4031122

Safety Information

signalword Danger
MSDS https://www.hit2lead.com/building-blocks/4031122
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart