CH4182077316 ChemBridge Corporation

2,6-dimethyl-1H-indole-3-carbaldehyde

  • MDL number MFCD06016187
  • Cas Number : 728024-59-7
  • Molecular Weight : 173.2111

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4031258

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4031258
Availability details for
Quantity
  • Availability
  • Add to Cart