CH4182077349 ChemBridge Corporation

1-(4-chlorophenyl)-1-(3-pyridinyl)methanamine

  • MDL number MFCD08144803
  • Cas Number : 883548-10-5
  • Molecular Weight : 218.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4031537

Safety Information

MSDS https://www.hit2lead.com/sds/4031537_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart