CH4182077351 ChemBridge Corporation

5-amino-6-methyl-1,3-dihydro-2H-benzimidazol-2-one

  • MDL number MFCD02684428
  • Cas Number : 67014-36-2
  • Molecular Weight : 163.1766

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4031542

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4031542
Availability details for
Quantity
  • Availability
  • Add to Cart