CH4182077388 ChemBridge Corporation

1-(4-methyl-1,2,3-thiadiazol-5-yl)methanamine

  • MDL number MFCD10001570
  • Cas Number : 790641-72-4
  • Molecular Weight : 129.1835

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4031769

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4031769
Availability details for
Quantity
  • Availability
  • Add to Cart