CH4182077409 ChemBridge Corporation

6-fluoro-1-methyl-1H-indole-3-carbaldehyde

  • MDL number MFCD07790764
  • Cas Number : 441715-93-1
  • Molecular Weight : 177.175

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4031991

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4031991
Availability details for
Quantity
  • Availability
  • Add to Cart