CH4182077415 ChemBridge Corporation

(4-[1,2,4]triazolo[3,4-b][1,3,4]thiadiazol-6-ylphenyl)amine

  • MDL number MFCD02249088
  • Cas Number : 797767-52-3
  • Molecular Weight : 217.2504

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4032022

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4032022
Availability details for
Quantity
  • Availability
  • Add to Cart