CH4182077423 ChemBridge Corporation

1-ethyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD02090832
  • Cas Number : 400755-44-4
  • Molecular Weight : 140.1399

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4029168

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4029168
Availability details for
Quantity
  • Availability
  • Add to Cart