CH4182077426 ChemBridge Corporation

(4-bromo-1H-pyrazol-1-yl)acetic acid

  • MDL number MFCD00159632
  • Cas Number : 82231-53-6
  • Molecular Weight : 205.0094

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4032060

Safety Information

MSDS https://www.hit2lead.com/sds/4032060_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart