CH4182077443 ChemBridge Corporation

3-(1H-imidazol-2-yl)pyridine

  • MDL number MFCD08458369
  • Cas Number : 13570-00-8
  • Molecular Weight : 145.1613

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4032611

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4032611
Availability details for
Quantity
  • Availability
  • Add to Cart