CH4182077448 ChemBridge Corporation

4,6-dimethyl-1H-indole-2-carboxylic acid

  • MDL number MFCD02664431
  • Cas Number : 383132-27-2
  • Molecular Weight : 189.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4032863

Safety Information

MSDS https://www.hit2lead.com/sds/4032863_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart