CH4182077465 ChemBridge Corporation

1-hydroxycyclohexanecarboxylic acid

  • MDL number MFCD00045477
  • Cas Number : 1123-28-0
  • Molecular Weight : 144.1684

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033358

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4033358
Availability details for
Quantity
  • Availability
  • Add to Cart