CH4182077469 ChemBridge Corporation

methyl 4-bromo-1-methyl-1H-pyrazole-5-carboxylate

  • MDL number MFCD01459850
  • Cas Number : 514816-42-3
  • Molecular Weight : 219.036

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033387

Safety Information

MSDS https://www.hit2lead.com/sds/4033387_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart