CH4182077481 ChemBridge Corporation

4-bromo-1-methyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD00463982
  • Cas Number : 84547-84-2
  • Molecular Weight : 205.0094

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033665

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4033665
Availability details for
Quantity
  • Availability
  • Add to Cart