CH4182077487 ChemBridge Corporation

2-chloro-6-fluoro-4-methylquinoline

  • MDL number MFCD03085779
  • Cas Number : 18529-12-9
  • Molecular Weight : 195.6207

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Assay 0.95
Storage temp. Room Temperature

Safety Information

msds https://www.hit2lead.com/building-blocks/4033831
Availability details for
Quantity
  • Availability
  • Add to Cart