CH4182077488 ChemBridge Corporation

2-(4-morpholinyl)-1H-benzimidazole

  • MDL number MFCD00569559
  • Cas Number : 31075-58-8
  • Molecular Weight : 203.2404

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033874

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4033874
Availability details for
Quantity
  • Availability
  • Add to Cart