CH4182077504 ChemBridge Corporation

1-(3-pyridinylmethyl)-4-piperidinecarboxylic acid

  • MDL number MFCD03371464
  • Cas Number : 887442-22-0
  • Molecular Weight : 220.2676

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4034188

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4034188
Availability details for
Quantity
  • Availability
  • Add to Cart