CH4182077551 ChemBridge Corporation

2-pyrazinylmalonaldehyde

  • MDL number MFCD00216533
  • Cas Number : 13481-00-0
  • Molecular Weight : 150.1347

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4034745

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4034745
Availability details for
Quantity
  • Availability
  • Add to Cart