CH4182077590 ChemBridge Corporation

2-(2-azabicyclo[2.2.1]hept-5-en-2-yl)ethanamine

  • MDL number MFCD09055376
  • Cas Number : 933750-04-0
  • Molecular Weight : 138.2102

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4101072

Safety Information

MSDS https://www.hit2lead.com/sds/4101072_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart