CH4182077614 ChemBridge Corporation

rac-(1R,2R,4R)-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid

  • MDL number MFCD08899816
  • Cas Number : 58001-99-3
  • Molecular Weight : 138.1638

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4101659

Safety Information

MSDS https://www.hit2lead.com/sds/4101659_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart