CH4182077618 ChemBridge Corporation

10-methoxy-3,4,5,6-tetrahydro-2H-1,5-benzoxazocine

  • MDL number MFCD09055380
  • Cas Number : 938459-13-3
  • Molecular Weight : 193.2423

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4101795

Safety Information

MSDS https://www.hit2lead.com/sds/4101795_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart