CH4182077629 ChemBridge Corporation

1-(1-morpholin-4-ylcyclopentyl)methanamine

  • MDL number MFCD08703639
  • Cas Number : 444666-61-9
  • Molecular Weight : 184.2786

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4102172

Safety Information

MSDS https://www.hit2lead.com/sds/4102172_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart