CH4182077630 ChemBridge Corporation

1-(1-piperidin-1-ylcyclopentyl)methanamine

  • MDL number MFCD09055382
  • Cas Number : 933701-05-4
  • Molecular Weight : 182.3058

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4102173

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4102173
Availability details for
Quantity
  • Availability
  • Add to Cart