CH4182077665 ChemBridge Corporation

3-methyl-1H-indole-2-carboxylic acid

  • MDL number MFCD01652241
  • Cas Number : 10590-73-5
  • Molecular Weight : 175.184

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4140536

Safety Information

MSDS https://www.hit2lead.com/sds/4140536_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart