CH4182077685 ChemBridge Corporation

N~3~-isopropyl-beta-alaninamide

  • MDL number MFCD10686616
  • Cas Number : 20101-89-7
  • Molecular Weight : 130.1882

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4029400

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4029400
Availability details for
Quantity
  • Availability
  • Add to Cart