CH4182077702 ChemBridge Corporation

3-(2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)propanoic acid

  • MDL number MFCD00454329
  • Cas Number : 75655-44-6
  • Molecular Weight : 206.198

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4401357

Safety Information

MSDS https://www.hit2lead.com/sds/4401357_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart