CH4182077719 ChemBridge Corporation

(5-methoxy-2-oxo-2,3-dihydro-1H-indol-3-yl)acetic acid

  • MDL number MFCD06738900
  • Cas Number : 885272-25-3
  • Molecular Weight : 221.2093

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4402028

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4402028
Availability details for
Quantity
  • Availability
  • Add to Cart