CH4182077725 ChemBridge Corporation

4-acetyl-3,4-dihydro-2H-1,4-benzoxazine-6-sulfonyl chloride

  • MDL number MFCD09972061
  • Cas Number : 1017791-37-5
  • Molecular Weight : 275.7087

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4029429

Safety Information

signalword Danger
MSDS https://www.hit2lead.com/building-blocks/4029429
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart