CH4182077762 ChemBridge Corporation

3-(3-methylbutoxy)benzaldehyde

  • MDL number MFCD09864622
  • Cas Number : 77422-25-4
  • Molecular Weight : 192.2542

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071441

Safety Information

MSDS https://www.hit2lead.com/building-blocks/9071441
Availability details for
Quantity
  • Availability
  • Add to Cart