CH4183975473 ChemBridge Corporation

1,4-dimethyl-1H-indole-3-carbaldehyde

  • MDL number MFCD11505328
  • Cas Number : 170489-16-4
  • Molecular Weight : 173.2111

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033745

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4033745
Availability details for
Quantity
  • Availability
  • Add to Cart