CH4183975567 ChemBridge Corporation

(4-sec-butylphenyl)boronic acid

  • MDL number MFCD02179456
  • Cas Number : 850568-56-8
  • Molecular Weight : 178.0359

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4030859

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/building-blocks/4030859
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart