CH4183975607 ChemBridge Corporation

4-bromo-1-ethyl-5-methyl-1H-pyrazole

  • MDL number MFCD11500994
  • Cas Number : 1171667-09-6
  • Molecular Weight : 189.0531

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4032178

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4032178
Availability details for
Quantity
  • Availability
  • Add to Cart