CH4183975619 ChemBridge Corporation

4-amino-N,N,3-trimethylbenzamide

  • MDL number MFCD09728471
  • Cas Number : 953739-92-9
  • Molecular Weight : 178.231

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4032444

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4032444
Availability details for
Quantity
  • Availability
  • Add to Cart