CH4183975626 ChemBridge Corporation

(2-ethyl-1H-imidazol-1-yl)acetic acid

  • MDL number MFCD07186426
  • Cas Number : 883539-33-1
  • Molecular Weight : 154.1665

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4013163

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4013163
Availability details for
Quantity
  • Availability
  • Add to Cart