CH4183975658 ChemBridge Corporation

1,7-dimethyl-1H-indole-3-carbaldehyde

  • MDL number MFCD11505321
  • Cas Number : 164353-61-1
  • Molecular Weight : 173.2111

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033240

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4033240
Availability details for
Quantity
  • Availability
  • Add to Cart