CH4183975667 ChemBridge Corporation

2-(1H-1,2,4-triazol-1-yl)benzoic acid

  • MDL number MFCD11156565
  • Cas Number : 138479-54-6
  • Molecular Weight : 189.1708

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033379

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4033379
Availability details for
Quantity
  • Availability
  • Add to Cart