CH4183975680 ChemBridge Corporation

1-(5-amino-2-methylphenyl)-2-pyrrolidinone

  • MDL number MFCD09045534
  • Cas Number : 69131-44-8
  • Molecular Weight : 190.2417

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033626

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4033626
Availability details for
Quantity
  • Availability
  • Add to Cart