CH4183975684 ChemBridge Corporation

3-methyl-2-thiophenecarbohydrazide

  • MDL number MFCD01993658
  • Cas Number : 350997-56-7
  • Molecular Weight : 156.2055

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033736

Safety Information

MSDS https://www.hit2lead.com/sds/4033736_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart