CH4183975742 ChemBridge Corporation

7-chloro-1H-indole-2-carboxylic acid

  • MDL number MFCD02664428
  • Cas Number : 28899-75-4
  • Molecular Weight : 195.6024

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4023476

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4023476
Availability details for
Quantity
  • Availability
  • Add to Cart